Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Modifier, 2.0% Mg (20,000 ppm Mg), Ricca Chemical
SDP

Catalog No. p-3251899 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Catalog No. MMMG20KN-50

Supplier: Ricca Chemical Company MMMG20KN50

Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Matrix Modifier
Packaging Natural Poly Bottle
Concentration or Composition Notes 2.0% Pd,20,000ppm Pd
CAS Min % 76.0, 16.09, 5.72
CAS Max % 79.0, 16.74, 5.96
CAS 13446-18-9, 7697-37-2
Physical Form Liquid
Color Colorless
Quantity 50 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.