Alfa Aesar™ Matrix oil, 20 centistoke hydrocarbon oil

Manufacturer: Alfa Aesar™ 44012LV

Quantity 1 gal.

 View more versions of this product

Catalog No. AA44012LV

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00147539
    Chemical Name or Material Matrix oil, 20 centistoke hydrocarbon oil

    Specifications

    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H350
    May cause cancer.
    Quantity 1 gal.
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00147539
    Health Hazard 1 Danger
    Chemical Name or Material Matrix oil, 20 centistoke hydrocarbon oil

    GHS H Statement
    H350
    May cause cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit