Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Matrix™ Pipette Tips

Catalog No. 14387180
Click to view available options
Volume (Metric):
1250 μL
Sterility:
Sterile
Format:
Rack

Choose from a comprehensive selection of these pipette tips designed together with pipetters to ensure a perfect balance between security of the seal and ease of application/ejection.

Choose from a comprehensive selection of pipette tips. Thermo Scientific™ Matrix Pipette Tips are supplied in innovative new Thermo Scientific™ Flo-Thru™ racks. Matrix pipetters and pipet tips are designed together to ensure that a perfect balance exists between security of the seal and ease of application/ejection. Manufactured under tight QC specifications, these standard tips rival special low-retention tips currently on the market after pipetting fluids such as dH2O, 50% Glycerol, Hybridized Buffer and 0.1% SDS.

Features:

  • Sealing rings on tips ensure confident, leak-free seal without banging tips on pipetter
  • Because the sealing rings compress, these tips also offer a secure seal on most other pipetters on the market
  • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives
  • Stabilization rings in racks hold tips steady when seating on pipetter
  • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips.
  • Finger notch on racks allows easy removal/insertion of refill trays.

Innovative Racks and Packaging:

  • Rigid rack design ensures solid, even tip seating; rack will not flex or bend when trying to apply tips.
  • Unique Thermo Scientific Flo-Thru rack individually supports tips to prevent them from wobbling when trying to seat them on the pipetter.
  • Hinge prevents sterile lid from coming in contact with lab bench.
  • Venting along the edge or around each tip improves heat flow during autoclaving; reduces condensation that can form on the tips.
  • Dual-position lid can be propped open to prevent vacuum deformation during autoclaving.
  • Sterile racks are sold in individual bags with quick-tear openings; each rack remains sterile until ready for use.
  • Each rack is labeled with product details including re-ordering information, description, volume range and lot number.
  • Racks are constructed of clean recycled materials and are made as small as possible to reduce waste.
  • Racks are marked with appropriate symbols to support recycling initiatives.
TRUSTED_SUSTAINABILITY

Specifications

Description Matrix 1250, Hinged
Volume (Metric) 1250 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 7.9cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
Format Rack
Tip Style Low Retention
For Use With (Equipment) Thermo Scientific Matrix Pipette
Product Line Matrix
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.