Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Matrix™ SepraSeal and DuraSeal Capping Systems
GREENER_CHOICE

Catalog No. 04100534
Change view
Click to view available options
Product Line:
Matrix™ DuraSeal
Matrix™ SepraSeal
Description:
Bulk Caps
Mats
Mats, Pre-Split
Color:
Blue
Gray
Green
Natural
Purple
Red
Yellow
Sterility:
Non-sterile
Sterile
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line Description Color Sterility
04-100-534 Matrix™ SepraSeal Mats Red Non-sterile
04-100-530 Matrix™ SepraSeal Mats Blue Non-sterile
04-100-531 Matrix™ SepraSeal Mats Green Non-sterile
04-100-532 Matrix™ SepraSeal Mats Gray Non-sterile
04-100-533 Matrix™ SepraSeal Mats Purple Non-sterile
04-100-535 Matrix™ SepraSeal Mats Yellow Non-sterile
04-100-537 Matrix™ SepraSeal Mats Blue Sterile
04-100-538 Matrix™ SepraSeal Mats Green Sterile
04-100-539 Matrix™ SepraSeal Mats Gray Sterile
04-100-540 Matrix™ SepraSeal Mats Purple Sterile
04-100-541 Matrix™ SepraSeal Mats Red Sterile
04-100-542 Matrix™ SepraSeal Mats Yellow Sterile
04-100-544 Matrix™ SepraSeal Mats, Pre-Split Blue Non-sterile
04-100-545 Matrix™ SepraSeal Mats, Pre-Split Green Non-sterile
04-100-546 Matrix™ SepraSeal Mats, Pre-Split Gray Non-sterile
04-100-547 Matrix™ SepraSeal Mats, Pre-Split Purple Non-sterile
04-100-548 Matrix™ SepraSeal Mats, Pre-Split Red Non-sterile
04-100-549 Matrix™ SepraSeal Mats, Pre-Split Yellow Non-sterile
04-100-551 Matrix™ SepraSeal Mats, Pre-Split Blue Sterile
04-100-552 Matrix™ SepraSeal Mats, Pre-Split Green Sterile
04-100-553 Matrix™ SepraSeal Mats, Pre-Split Gray Sterile
04-100-554 Matrix™ SepraSeal Mats, Pre-Split Purple Sterile
04-100-555 Matrix™ SepraSeal Mats, Pre-Split Red Sterile
04-100-556 Matrix™ SepraSeal Mats, Pre-Split Yellow Sterile
04-100-543 Matrix™ SepraSeal Mats, Pre-Split Natural Non-sterile
04-100-558 Matrix™ DuraSeal Bulk Caps Natural Non-sterile
04-100-529 Matrix™ SepraSeal Mats Natural Non-sterile
04-100-550 Matrix™ SepraSeal Mats, Pre-Split Natural Sterile
04-100-536 Matrix™ SepraSeal Mats Natural Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
This item is not returnable. View return policy
Catalog No. 04-100-534 Supplier Thermo Scientific™ Supplier No. 4463RED
Add to Cart
Add to Cart
This item is not returnable. View return policy

Accommodate a range of application and storage requirements. These flexible capping options allow automated piercing, visual identification and sterility preference.

Accommodate a range of application and storage requirements. Thermo Scientific™ Matrix™ SepraSeal and DuraSeal tube sealing solutions complement the innovative line of Thermo Scientific Matrix 2D and non-2D coded storage tubes. Available in a range of formats to allow automated piercing, visual identification and sterility preference to meet your individual storage needs.

Sample Access Options

  • Designed for storage down to -20°C, Matrix SepraSeal mats are available in sterile, non-sterile, solid, or pre-split, and a range of colors for quick visual sample identification
  • Solid seals can be removed for sample access with the Thermo Scientific Matrix SepraSeal cap removal tool, or can be pierced once
  • For applications involving repetitive sample access, a pre-split Matrix SepraSeal can be pierced by automated liquid handling equipment, requiring very little pressure, while maintaining seal integrity with self-sealing closure
  • For applications requiring storage down to -80°C, permanently installed DuraSeals are an ultra-robust SepraSeal option—can also be used for repetitive piercing procedures
  • The Matirx DuraSeals Pre-Split V-Shaped design reliably and accurately guides automated liquid handling needles into the same location over multiple piercings—self-sealing design maintains sample integrity and prevents loss during storage

Application Flexibility

  • Elastomer ensures sealing performance will not degrade, even after multiple cap piercings
  • The Matrix SepraSeal tube sealing solution offers the flexibility of individual tube capping while also providing the ability to apply 96 caps at once
  • Supplied in mats of 96 caps each, sealing is secure and consistent every time when applied with the Thermo Scientific SuperSealer
  • After the SuperSealer applies the mat, simply pull up the backing sheet to reveal 96 individually sealed tubes—for applications where multiple piercings are required
  • The Matrix DuraSeal capping system is available in bulk or pre-installed in a wide array of our storage tubes
TRUSTED_SUSTAINABILITY

Specifications

Product Line Matrix™ SepraSeal
Color Red
Description Mats
No. per Case 10 Mats of 96 Caps/Cs.
Sterility Non-sterile
Product Type Croygenic Tube Closure
What are the main advantages of the Thermo Scientific 2D barcoded storage system?

Here are some advantages of the 2D barcoded storage system:
- Less time needed for labeling, sample recording, and retrieval
- Maximize freezer utilization while keeping storage space organised
- Ensures a safe, permanent ID of a sample
- Enables multivial capping and multichannel filling
- Individualized samples, eliminating multiple freeze/thaw cycles

Which Thermo Scientific 2D barcoded storage tubes should I use for my application?

Please view the following information regarding the usage of Thermo Scientific 2D barcoded storage tubes:
Tube Type; Typical Storage Temperature; Application(s); Special Feature(s)
Sepraseal; -20 degrees C; Compound storage; Can easily be removed for sample access
Presplit Sepraseal; -20 degrees C; Compound storage; Up to 200 piercings
Duraseal; -20 degrees; Compound storage; Safe seal for multiple piercings
Matrix ScrewTop; -80 degrees C; Blood components, DNA, Bacteria; With the correct torque, there is very little evaporation at -80 C, disposable pipette tips can be used
Nunc Screwcap; -176 degrees C; Mammalian cells; Minimal height, tubes can be removed with cryo gloves

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.