Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Matrix™ TallTip™ Extended Length, Racked Pipette Tips
Click to view available options
Volume (Metric):
1250 μL
Sterility:
Non-sterile
Format:
Rack
Description
Maximize reach with Thermo Scientific™ Matrix™ TallTip™ Extended Length Pipette Tips, which allow access to the bottom of test tubes, reagent bottles, flasks and other vessels without touching the barrel of the pipetter against the side of the tube. Nonbeveled, narrow tip ends provide cleaner touch-off of sidewalls for greater precision
Benefits:
- Prevent cross-contamination from vessel walls
- Allow the use of large volume tips in small spaces resulting in less liquid displacement
- 102 mm maximum length offers safe access to 100 mm test tubes
- Thermo Scientific™ Flo-Thru™ rack reduces tip wobble and improves airflow while autoclaving
- Universal fit works with most major brands of pipetters
Matrix Pipet Tip Features
- Sealing rings on tips ensure a confident seal without banging tips on pipetter
- Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives
- Stabilization rings in racks hold tips steady when seated on a pipetter
- Rigid rack design ensures solid, even tip seating
- Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips
- Hinged lid prevents sterile lid from coming in contact with lab bench
- Finger notch on racks allows easy removal/insert of refill trays
- Snap-and-tear bags make it easy to open each individually wrapped sterile rack
- Well-labeled racks simplify record keeping, tip identification and re-ordering
- Environmentally sensitive reduced rack size reduces waste
- Racks utilize recycled material and are marked with appropriate symbols to support recycling initiatives

Specifications
Specifications
Volume (Metric) | 1250 μL |
Filtered | Non-filtered |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Format | Rack |
Tip Style | Extended Length |
For Use With (Equipment) | Universal Fit Pipettor |
Product Line | Matrix TallTip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction