Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

All About Matter Learning Guide
SCI_ED

Catalog No. s13897 Shop All NewPath Learning Products
Click to view available options
Format:
36-page workbook

Catalog No. S13897

Supplier: NewPath Learning 143651

Only null left
Add to Cart
Add to Cart

Use engaging graphics and inquiry-based activities to enhance student comprehension on standards-based concepts on Matter and its Properties

Full-color, 36-page guide features self-directed readings, easy-to-follow illustrated explanations; guiding questions; inquiry-based activities; a lab investigation; key vocabulary review; and assessment review questions along with a post-test

Topics covered:

  • What is Matter?
  • Describing Matter/Properties of Matter
  • Physical Properties of Matter
  • Measuring Matter
  • States of Matter
  • Changes to Matter (Gas, Liquid, Solid)
  • Mixtures and Solutions
  • Chemical Change

Specifications

Product Type Learning Guide
Format 36-page workbook
For Use With (Application) Enhance student comprehension on standards-based concepts on matter and its properties
Certifications/Compliance Meets Academic Standard for California, Florida, National, New York, Texas and Next Generation Science Standards
Quantity 10/Pk.
Grade Level Elementary
Grade 3 to 5
Subject Physical Science
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.