Learn More
Invitrogen™ Matuzumab Chimeric Recombinant Rabbit Monoclonal Antibody (Matuzumab)

Description
Specificity: This antibody binds the extracellular domain III of the human EGFR and does not cross react with murine EGFR. It was also reported that residues S460/G461 in EGFR domain III are essential components of the epitope. This antibody binds at a site distinct from the EGF binding site, preventing conformational re-arrangement required for dimerization, a process essential for high-affinitiy ligand binding. This antibody can only recognize the active form of EGFR. The antibody binds to both glyco- and aglyco receptor forms.
Specifications
Specifications
| Antigen | Matuzumab Chimeric |
| Applications | ELISA, Flow Cytometry, Functional Assay, Inhibition Assays, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | Matuzumab |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | Cd8b1 |
| Gene Accession No. | P10300 |
| Gene Alias | EMD-72000; H425 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.