Learn More
Invitrogen™ Matuzumab Recombinant Human Monoclonal Antibody (Matuzumab)
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547997
Description
Specificity: This antibody binds the extracellular domain III of the human EGFR and does not cross react with murine EGFR. It was also reported that residues S460/G461 in EGFR domain III are essential components of the epitope. This antibody binds at a site distinct from the EGF binding site, preventing conformational re-arrangement required for dimerization, a process essential for high-affinitiy ligand binding. This antibody can only recognize the active form of EGFR. The antibody binds to both glyco- and aglyco receptor forms.
Matuzumab is a humanized monoclonal antibody used in cancer treatment. It has a high affinity for EGFR, frequently associated with the growth of blood vessels in malignancy, facilitating tumor growth and survival.
Specifications
Matuzumab | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P16871 | |
Human | |
Protein A | |
RUO | |
3575 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Functional Assay, Inhibition Assays, Western Blot, Immunocytochemistry | |
Matuzumab | |
Unconjugated | |
Il7r | |
EMD-72000; H425 | |
Domain III of the extracellular domain of human EGRF. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.