Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAU2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570088 Supplier Invitrogen™ Supplier No. PA570088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Plays an important role in the loading of the cohesin complex on to DNA. Forms a heterodimeric complex (also known as cohesin loading complex) with NIPBL/SCC2 which mediates the loading of the cohesin complex onto chromatin (PubMed:28167679, PubMed:22628566). Plays a role in sister chromatid cohesion and normal progression through prometaphase (PubMed:16802858, PubMed:16682347). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAU2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAU2
Gene Accession No. Q9Y6X3
Gene Alias 9130404D08Rik; A930019L04Rik; C77863; C79014; Cohesin loading complex subunit SCC4 homolog; KIAA0892; Mau2; Mau-2; MAU2 chromatid cohesion factor homolog; MAU2 chromatid cohesion factor homolog (C. elegans); MAU2 sister chromatid cohesion factor; MAU2, sister chromatid cohesion factor; MAU2L; mKIAA0892; protein MAU-2; RGD1308759; Scc4; Scc4-like protein; sister chromatid cohesion 4
Gene Symbols MAU2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MAU2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23383
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.