Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAU2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566290
Description
Immunogen sequence: HWLPKEHMCVL VYLVTVMHSM QAGYLEKAQK YTDKALMQLE KLKMLDCSPI LSSFQVILLE HIIMCRLVTG HKATALQEIS QVCQLCQQSP RLFSNHAAQ Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Plays an important role in the loading of the cohesin complex on to DNA. Forms a heterodimeric complex (also known as cohesin loading complex) with NIPBL/SCC2 which mediates the loading of the cohesin complex onto chromatin (PubMed:28167679, PubMed:22628566). Plays a role in sister chromatid cohesion and normal progression through prometaphase (PubMed:16802858, PubMed:16682347). [UniProt].
Specifications
MAU2 | |
Polyclonal | |
Unconjugated | |
MAU2 | |
9130404D08Rik; A930019L04Rik; C77863; C79014; Cohesin loading complex subunit SCC4 homolog; KIAA0892; Mau2; Mau-2; MAU2 chromatid cohesion factor homolog; MAU2 chromatid cohesion factor homolog (C. elegans); MAU2 sister chromatid cohesion factor; MAU2, sister chromatid cohesion factor; MAU2L; mKIAA0892; protein MAU-2; RGD1308759; Scc4; Scc4-like protein; sister chromatid cohesion 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23383 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6X3 | |
MAU2 | |
Recombinant Human MAU2. Recombinant protein control fragment (Product #RP-105142). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction