Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAVS Antibody (58N3B6), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP222380UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222380UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222380UV Supplier Novus Biologicals Supplier No. NBP222380UV
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MAVS
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 58N3B6
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. AAH44952.1
Gene Alias CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051
Gene Symbols MAVS
Host Species Mouse
Immunogen Amino acids 93-276 of human MAVS (isoform 1) were used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 57506
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.