Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAVS Antibody (58N3E1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen MAVS
Clone 58N3E1
Concentration 0.5 mg/mL
Dilution Western Blot 0.1-2 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP222378SS
SDP
View Documents
Novus Biologicals
NBP222378SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP222378
SDP
View Documents
Novus Biologicals
NBP222378
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Western Blot.
Specifications

Specifications

MAVS
0.5 mg/mL
Western Blot
Unconjugated
Mouse
Immune System Diseases, Immunology, Protein Kinase
AAH44952.1
57506
Within the range of amino acids 93-298 of human MAVS (isoform 1) were used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
58N3E1
Western Blot 0.1-2 ug/ml
Monoclonal
Purified
RUO
Human
CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051
MAVS
IgG2b κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.