Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAVS Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177275
Description
MAVS Polyclonal specifically detects MAVS in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Chromatin Immunoprecipitation (ChIP).Specifications
MAVS | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 0.5-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml, Chromatin Immunoprecipitation (ChIP) | |
NP_065797 | |
MAVS | |
Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human VISA. The immunogen is located within the first 50 amino acids of VISA. Amino Acid Squence: CLTARDQDRLRAT | |
0.1 mg | |
Immune System Diseases, Immunology, Protein Kinase | |
57506 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction