Learn More
Invitrogen™ MAVS Polyclonal Antibody

Description
A suggested positive control is rat brain tissue lysate. PA5-20349 can be used with blocking peptide PEP-0469.
Specifications
Specifications
| Antigen | MAVS |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MAVS |
| Gene Accession No. | Q66HG9, Q7Z434, Q8VCF0 |
| Gene Alias | CARD adapter inducing interferon beta; CARD adaptor inducing IFN-beta; Cardif; D430028G21Rik; IFN-B promoter stimulator 1; IFN-beta promoter stimulator-1; interferon beta promoter stimulator protein 1; interferon-beta promoter stimulator protein 1; IPS1; IPS-1; KIAA1271; MAVS; mitochondrial antiviral signaling protein; mitochondrial anti-viral signaling protein; mitochondrial antiviral-signaling protein; Putative NF-kappa-B-activating protein 031N; virus-induced signaling adapter; virus-induced signaling adaptor; virus-induced-signaling adapter; VISA |
| Gene Symbols | MAVS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.