Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAVS Recombinant Rabbit Monoclonal Antibody (PSH0-30), Invitrogen™

Catalog No. FEMA550801
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550801 Supplier Thermo Scientific Supplier No. MA550801

Rabbit Recombinant Monoclonal Antibody

Positive control: MCF7 cell lysate, C2C12 cell lysate, human colon tissue, human endometrium tissue, A431 cell. Predicted band size: 56 kDa Subcellular Location: Membrane, Mitochondrion, Mitochondrion outer membrane, Peroxisome.

Two distinct signaling pathways activate the host innate immunity against viral infection. One pathway is reliant on members of the Toll-like receptor (TLR) family while the other uses the RNA helicase RIG-I as a receptor for intracellular viral double-stranded RNA as a trigger for the immune response. MAVS is a mitochondrial membrane protein that was identified as a critical component in the IFN beta signaling pathways that recruits IRF-3 to RIG-I, leading to its activation and that of NF-kappa-B. MAVS is also thought to interact with other components of the innate immune pathway such as the TLR adapter protein TRIF, TRAF2 and TRAF6. MAVS also interacts with the IKK-alpha, IKK-beta and IKK-iota kinases through its C-terminal region. Cleavage of this region by the Hepatitis C virus (HCV) protease allows HCV to escape the host immune system. Multiple isoforms of MAVS are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAVS
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PSH0-30
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MAVS
Gene Accession No. Q66HG9, Q7Z434, Q8VCF0
Gene Alias CARD adapter inducing interferon beta, CARD adaptor inducing IFN-beta, Cardif, D430028G21Rik, IFN-B promoter stimulator 1, IFN-beta promoter stimulator-1, interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS1, IPS-1, KIAA1271, MAVS, mitochondrial antiviral signaling protein, mitochondrial anti-viral signaling protein, mitochondrial antiviral-signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter, virus-induced signaling adaptor, virus-induced-signaling adapter, VISA
Gene Symbols MAVS
Host Species Rabbit
Immunogen Synthetic peptide within human MAVS aa 2-50/540.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228607, 311430, 57506
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.