Learn More
MAVS/VISA Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300782AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
Two distinct signaling pathways activate the host innate immunity against viral infection. One pathway is reliant on members of the Toll-like receptor (TLR) family while the other uses the RNA helicase RIG-I as a receptor for intracellular viral double-stranded RNA as a trigger for the immune response. VISA is a mitochondrial membrane protein that was identified as a critical component in the IFN-b signaling pathways that recruits IRF-3 to RIG-I, leading to its activation and that of NF-kappa-B. VISA is also thought to interact with other components of the innate immune pathway such as the TLR adapter protein TRIF, TRAF2 and TRAF6. VISA also interacts with the IKK-a, IKK-b and IKK-iota kinases through its C-terminal region. Cleavage of this region by the Hepatitis C virus (HCV) protease allows HCV to escape the host immune system. At least three isoforms of VISA are known to exist.Specifications
| MAVS/VISA | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q7Z434 | |
| MAVS | |
| Between 150 and 200 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| MAVS | |
| CARD adapter inducing interferon beta; CARD adapter inducing interferon-beta; CARD adaptor inducing IFN-beta; IFN-B promoter stimulator 1; interferon beta promoter stimulator protein 1; interferon-beta promoter stimulator protein 1; mitochondrial anti-viral signaling protein; mitochondrial antiviral-signaling protein; putative NF-kappa-B-activating protein 031N; virus-induced signaling adapter variant 1b; virus-induced signaling adaptor variant 1a; virus-induced-signaling adapter | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57506 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.