Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Max Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300047IR
Description
Max Polyclonal specifically detects Max in Human samples. It is validated for Western Blot, ELISA.Specifications
Max | |
Polyclonal | |
Western Blot, ELISA | |
BHLHD4, bHLHd4MGC11225, bHLHd5, bHLHd6, bHLHd7, bHLHd8, Class D basic helix-loop-helix protein 4, helix-loop-helix zipper protein, MAX protein, MGC10775, MGC18164, MGC34679, MGC36767, MYC associated factor X, Myc-associated factor X, orf1, protein max | |
Produced in rabbits immunized with purified, recombinant Max (NP_002373; Met1-Ser160). | |
0.1 mL | |
Apoptosis, Phospho Specific | |
4149 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction