Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAX binding protein Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204052G
Description
MAX binding protein Polyclonal antibody specifically detects MAX binding protein in Human samples. It is validated for Western BlotSpecifications
MAX binding protein | |
Polyclonal | |
Western Blot | |
BHLHD3, bHLHd3Max-interacting protein, Class D basic helix-loop-helix protein 3, MAD6, MAX binding protein, MXD6, Myc antagonist MNT, Protein ROX, ROXmax-binding protein MNT | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human MAX Binding Protein using the numbering given in entry NP_064706.1 (GeneID 4335). | |
0.1 mL | |
Cell Cycle and Replication | |
4335 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction