Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MAX™ Enteric Viral Panel

Catalog No. b443985 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
24/Pk.
This item is not returnable. View return policy

Catalog No. B443985

Supplier: BD 443985

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

BD™ MAX™ Enteric Viral Panel designed to detect the viral cause of infectious diarrhea symptoms.

  • Completes the suite of enteric panels that are available in the BD™ MAX™ System
  • Capable of running FDA cleared and open-system assays
  • Targeted panel approach ideal for syndromic testing
  • Detects norovirus, rotavirus, adenovirus, human astrovirus, and sapovirus
  • Results are provided in approximately 3 hours
  • Allows clinicians to more quickly understand the cause of a patient’s illness
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 25°C
Sample Type Unpreserved Soft to Diarrheal Stool or Cary-Blair Preserved Stool
Packaging Type 24/Pk.
Quantity 24/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.