Learn More
BD MAX™ Extended Enteric Bacterial Panel
Catalog No. b443812 Shop All BD Diagnostic Systems Products
Click to view available options
Test Time:
≈3.5 hr.
This item is not returnable. View return policy
Catalog No. B443812
Supplier: BD 443812
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
By adding the BD MAX™ Extended Enteric Bacterial Panel to your laboratory’s stool testing, over 95% of bacteria causing infectious gastroenteritis can be detected when run in conjunction with BD MAX™ Enteric Bacterial Panel.
The BD MAX™ Extended Enteric Bacterial Panel performed on the BD MAX System, is an automated in vitro diagnostic test for the direct qualitative detection and differentiation of enteric bacterial pathogens. It is used in conjunction with the BD MAX™ Enteric Bacterial Panel as an optional Master Mix addition to the 4 snap Unitized Reagent Strip. The BD MAX Extended Enteric Bacterial Panel detects nucleic acids from: Plesiomonas shigelloides, Vibrio (V. vulnificus, V. parahaemolyticus, and V. cholerae), Enterotoxigenic Escherichia coli (ETEC) heat-labile enterotoxin (LT)/ heat-stable enterotoxin (ST) genes, Yersina enterocolitica. Testing is performed on unpreserved soft to diarrheal stool specimens or Cary-Blair preserved stool specimens from symptomatic patients with suspected acute gastroenteritis, enteritis or colitis. The test is performed directly on the specimen, utilizing real-time polymerase chain reaction (PCR) for the amplification of relevant gene target DNA. The test utilizes fluorogenic gene-specific hybridization probes for the detection of the amplified DNA.  This test is intended for use, in conjunction with clinical presentation, laboratory findings, and epidemiological information, as an aid in the differential diagnosis of Plesiomonas shigelloides, Vibrio (V. vulnificus, V. parahaemolyticus, and V. cholerae) Enterotoxigenic Escherichia coli (ETEC) LT/ST and Yersinia enterocolitica infections. Results of this test should not be used as the sole basis for diagnosis, treatment, or other patient management decisions. Positive results do not rule out co-infection with other organisms that are not detected by this test, and may not be the sole or definitive cause of patient illness. Negative results in the setting of clinical illness compatible with gastroenteritis may be due to infection by pathogens that are not detected by this test or non-infectious causes such as ulcerative colitis, irritable bowel syndrome, or Crohn's disease.
TRUSTED_SUSTAINABILITY
Specifications
Sample Type Stool
Includes BD MAX™ Extended Enteric Bacterial Panel Master Mix (24)
Test Time ≈3.5 hr.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.