Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAX Monoclonal Antibody (01)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529411
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MAX/MYC associated factor X.
The protein encoded by this gene is a member of the basic helix-loop-helix leucine zipper (bHLHZ) family of transcription factors. It is able to form homodimers and heterodimers with other family members, which include Mad, Mxi1 and Myc. Myc is an oncoprotein implicated in cell proliferation, differentiation and apoptosis. The homodimers and heterodimers compete for a common DNA target site (the E box) and rearrangement among these dimer forms provides a complex system of transcriptional regulation. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature for some of them is unknown.
Specifications
MAX | |
Monoclonal | |
5 mg/mL | |
PBS with no preservative | |
P61244 | |
MAX | |
Recombinant Human MAX/MYC associated factor X protein (Met1-Ser160). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
01 | |
Unconjugated | |
MAX | |
AA960152; AI875693; bHLHd4; bHLHd5; bHLHd6; bHLHd7; bHLHd8; Class D basic helix-loop-helix protein 4; Delta-Max; Max; Max protein; MGC10775; MGC11225; MGC18164; MGC34679; MGC36767; MYC associated factor X; myc-associated factor X; Myc-binding novel HLH/LZ protein; Myn; orf1; Protein max; protein myn | |
Mouse | |
Protein A | |
RUO | |
4149 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction