Learn More
BD™ MAX™ UVE Specimen Collection Kit
Catalog No. B443376 Shop All BD Diagnostic Systems Products
Click to view available options
:
100 kits per shelfpack; each kit contains 1 BD MAX™ Specimen Collection Swab, 1 BD MAX™ UVE Sample Buffer Tube, 1 Graduated Transfer Pipette
This item is not returnable. View return policy
Catalog No. B443376
Supplier: BD™ 443376
This item is not returnable. View return policy
The BD MAX™ UVE Specimen Collection Kit is intended to be used in clinical settings according to the instructions provided for collection and transport of vaginal and endocervical swab specimens, in addition to the preservation of urine from males and females.
The BD MAX™ UVE Specimen Collection Kit is intended to be used in clinical settings according to the instructions provided for collection and transport of vaginal and endocervical swab specimens, in addition to the preservation of urine from males and females. This transport system is used for testing with the BD MAX products.
TRUSTED_SUSTAINABILITY
Specifications
Type Specimen Collection
Includes 100 kits per shelfpack; each kit contains 1 BD MAX™ Specimen Collection Swab, 1 BD MAX™ UVE Sample Buffer Tube, 1 Graduated Transfer Pipette
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.