Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maxi Potassium channel beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133484
Description
Maxi Potassium channel beta Polyclonal specifically detects Maxi Potassium channel beta in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Maxi Potassium channel beta | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
KCNMB1 | |
Recombinant protein encompassing a sequence within the center region of human KCNMB1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1.0mg/mL | |
Western Blot 1:1000-1:10000 | |
BK channel beta subunit, BK channel subunit beta-1, BKbeta, BKbeta1, Calcium-activated potassium channel subunit beta, calcium-activated potassium channel subunit beta-1, Calcium-activated potassium channel, subfamily M subunit beta-1, Charybdotoxin receptor subunit beta-1, Hbeta1, hslo-beta, K(VCA)beta, k(VCA)beta-1, large conductance Ca2+-activated K+ channel beta 1 subunit, Maxi K channel beta subunit, Maxi K channel subunit beta-1, potassium large conductance calcium-activated channel, subfamily M, beta member1, Slo-beta, Slo-beta-1 | |
Rabbit | |
22 kDa | |
100 μL | |
Potassium Channels | |
3779 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction