Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiDry™ Ultra Lightweight 3/4 Nitrile Coated Gloves with Non-Slip Grip

Catalog No. 19148461 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-461 Small
19-148-463 Large
19-148-464 X-Large
19-148-465 2X-Large
19-148-462 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-148-461 Supplier PIP™ Supplier No. 56425/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

MaxiDry™ oil-repellant gloves feature an ultra thin design to provide flexibility and dexterity. Foam nitrile coating is compatible with light oils to enhance grip capabilities.

  • Seamless string knit shell is comfortable and flexible
  • Foam nitrile coating is compatible with light oils for enhanced grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Increased resistance to oils via ATG's LiquiTech™ technology platform
  • Synthetic coating combined with an industry leading super light seamless knit liner provides comfort, sensitivity, fit, and dexterity
  • Locking Cuff ensures the glove doesn't slip on the hand
  • An exclusive elasticated support in the wrist area provides a locking effect for a close and snug fit
  • Micro-cup non-slip grip finish allows for a controlled and superior grip in oily and wet applications
  • Non-slip grip is only applied where it is actually needed (palm area) to ensure flexibility isn't compromised
  • All ingredients used in the manufacturing and construction of this product are compliant with the REACH legislation
  • Gloves are laundered prior to final packaging it clean and safe for use
  • Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile MicroFoam
Color Black, Purple
Exterior Finish Fully Textured
Hand Specific
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size Small
Style Standard
For Use With (Application) Assembly, Precision Handling, and General Maintenance
Glove Cuff Style Knit
Series 56-425
Yarn Gauge 15
Length (English) 8.7 in.
Length (Metric) 22 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.