Learn More
PIP™ MaxiDry™ Zero™ Double-Dipped MicroFoam Nitrile-Coated Gloves with Thermal Lining
Shop All Protective Industrial Products Inc. ProductsDescription
- Seamless construction increases comfort and breathability
- MicroFoam Nitrile coating is compatible with light oils to enhance grip and abrasion resistance
- Knit Wrist helps prevent dirt and debris from entering the glove
- Liquid repellence keeps moisture away from the skin to ensure hands remain warm
- High degree of thermal insulation without sacrificing flexibility, dexterity, weight, and comfort
- Gloves help to prevent the effects of wind chills for outdoor applications
- Coating remains flexible even in extremely cold environments
- Optimized grip delivered though the ultra soft coating to ensure maximum flex and contact with the items being handled
- Insulation liner is bonded away from flex and movement points of the hand (i.e. knuckles) to ensure comfort
- All ingredients used in manufacturing and construction are compliant with the REACH legislation
- Gloves are laundered prior to final packaging to ensure that they are clean and safe for use
- The Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
- The Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product
- Complies with FDA food handling requirements 21 CFR, Part 177
- Silicone-free
Specifications
Specifications
| Certifications/Compliance | FDA 21 CFR 177.2600 |
| Coating Material | Nitrile |
| Color | Purple |
| Exterior Finish | Fully Textured |
| Hand | Specific |
| Material | Nylon Lycra |
| Product Type | Cold-resistant Gloves |
| Size | Medium |
| For Use With (Application) | Outdoor Construction/Maintenance, Agriculture, Cold Food Storage, and Chilled Distribution |
| Glove Cuff Style | Knit |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
