Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Cut™ Engineered Yarn Gloves, Nitrile Coated MicroFoam Grip

Catalog No. 19160393 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Blue
Brown
Gray
Green
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-160-393 Large Brown
19-160-390 X-Small Blue
19-160-391 Small White
19-160-392 Medium Green
19-160-394 X-Large Yellow
19-160-395 2X-Large Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-160-393 Supplier PIP™ Supplier No. 348743/L
Add to Cart
Add to Cart

High-performance yarns and fibers offer cut protection, and black micro-foam nitrile coating assures grip for precision handling in dry environments.

  • High performance yarns and fibers developed using CutTech technology offers cut protection
  • Seamless knit engineered yarn
  • Proprietary black micro-foam nitrile coating offers 360° breathability
  • Reinforcement between thumb and forefinger, improves cut resistance and extends glove life
  • Micro-cup finish for optimized grip
  • Minimal penetration (strike through) leaving only soft liner resting against skin
  • Component material complies with Federal regulations for food contact, 21CFR, 170-199
  • Continuous knit cuff
  • EN Cut Level 3
  • Silicone free
  • Green shell

Applications:

DIY, finishing and inspection, handling of small oily parts and components, wiring operations.

Specifications

Certifications/Compliance FDA 21CFR, 170-199
Certifications/Compliance (Europe) CE
Construction Seamless Knit
Coating Material Nitrile Foam
Color Green, Black
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Engineered Yarn
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4331
For Use With (Application) Finishing and inspection, handling of oily parts and components, assembly, maintenance, logistics and warehousing, wiring operations
Glove Cuff Color Brown
Glove Cuff Style Continuous Knit
Model 34-8743/L
Series MaxiFlex™ Cut™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.