Learn More
3M™ Maxim™ 2X2 Safety Goggles
Versatile design for tacticle use
Specifications
Certifications/Compliance ANSI Z87.1-2003
Product Type Safety goggles
For Use With (Application) Provide protection against hazards such as dust and chemical splash
Frame Color Black
No. per Case 10/Cs.
View More Specs
Products 1
Catalog Number Mfr. No. Lens Tint No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint No. per Case Price Quantity & Availability  
19-060-0424
View Documents
3M™
7000127566
Gray 10/Cs. N/A N/A N/A
Description
Description
  • 3M Catalog# 40686-00000-10 , 3M Catalog# 40687-00000-10
Compliance
Meets the High Impact Requirements of ANSI Z87.1-2003.
Specifications
Specifications
ANSI Z87.1-2003
Provide protection against hazards such as dust and chemical splash
10/Cs.
Safety goggles
Black
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.