Learn More
3M™ Maxim™ Eyewear
Contemporary, athletic styling
Specifications
Lens Material Polycarbonate
View More Specs
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Products 1
Catalog Number Mfr. No. dimLensCoating Lens Tint Temple Style Price Quantity & Availability  
Catalog Number Mfr. No. dimLensCoating Lens Tint Temple Style Price Quantity & Availability  
19-121-575
View Documents
3M™
7010045586
Clear Adjustable Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
Feauture polycarbonate lenses that absorb 99.9% UV.
  • Three-position pantascopic lens angle adjustment
  • Dual-injected cushioned browguard
  • Soft nose adjustable nose bridge
  • Adjustable temple lengths
  • Ventilation channel helps to prevent fogging
  • 3M Catalog# 13250-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Specifications
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.