Learn More
Thermo Scientific™ Maxima First Strand cDNA Synthesis Kit for RT-qPCR, with dsDNase PROMO
Available with or without dsDNase, is a convenient system optimized for cDNA synthesis in two-step quantitative RT-PCR (RT-qPCR) applications.
Supplier: Thermo Scientific™ K1672
Description
The Maxima First Strand cDNA Synthesis Kit for RT-qPCR is capable of reproducible cDNA synthesis from a wide range of total RNA amounts (1 pg to 5 μg) at elevated temperatures (42–65°C). The synthesis reaction can be completed in 15–30 minutes. Components of the Maxima First Strand cDNA Synthesis Kit for RT-qPCR are pre-mixed to save time and to reduce the possibility of pipetting errors.
The Maxima First Strand cDNA Synthesis Kit for RT-qPCR with dsDNase also contains a novel double-strand specific DNase engineered to remove contaminating genomic DNA from RNA preps in two minutes without damage to quality or quantity of RNA. Highly specific activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved and dsDNase treated RNA can be directly added to reverse transcription.
Features of the Maxima First Strand cDNA Synthesis Kit for RT-qPCR include:
- High yields of full length cDNA up to 20 kb
- Efficient cDNA synthesis at wide temperature range (42–65°C)
- Increased synthesis rate—complete cDNA synthesis in 15–30 minutes
- High sensitivity and specificity
- Integrated gDNA removal step with dsDNase kit
Maxima First Strand cDNA Synthesis Kit with dsDNase contains:
- Maxima Enzyme Mix
- 5X Reaction Mix
- Nuclease-free water
Kit with dsDNase also contains
- dsDNase
- 10X dsDNase Buffer
Applications
- 2-step RT-PCR
- 2-step RT-qPCR
Additional information about reaction components
The Maxima Enzyme Mix contains Maxima Reverse Transcriptase and RiboLock RNase Inhibitor. The recombinant RiboLock RNase Inhibitor effectively protects RNA templates from degradation by RNases A, B, and C at temperatures up to 55°C.
5X Reaction Mix contains the remaining reaction components: reaction buffer, dNTPs, oligo(dT)18 and random hexamer primers. Nuclease-free water is provided for reaction set-up and dilution of sample RNA.
The absence of endo- and exo-deoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.
Specifications
Maxima Enzyme Mix (Maxima Reverse Transcriptase and RiboLock RNase Inhibitor), 5X Reaction Mix, dsDNase, 10X dsDNase Buffer, Nuclease-free Water, Store at -20°C | |
High | |
Reverse Transcription | |
1 ea. | |
Yes | |
Real Time PCR (qPCR) | |
200 Reactions | |
Reverse Transcription | |
RNA |
Kit | |
30 min. | |
50°C to 55°C | |
Maxima | |
Dry Ice | |
First-Strand cDNA | |
Separate Components | |
Up to 20 kb | |
Kit with dsDNase |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.