Learn More
Thermo Scientific™ Maxima H Minus Double-Stranded cDNA Synthesis Kit
The Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA.
$873.00 - $7800.00
Specifications
Product Line | Maxima |
---|---|
Product Type | cDNA Synthesis Kit |
Reverse Transcriptase | Maxima H Minus |
For Use With (Application) | Cloning |
Final Product Type | Double Stranded cDNA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FERK2562
|
Thermo Scientific™
K2562 |
50 Reactions |
Each for $3,630.00
|
|
|||||
FERK2563
|
Thermo Scientific™
K2563 |
200 Reactions |
Each for $7,800.00
|
|
|||||
FERK2561
|
Thermo Scientific™
K2561 |
10 Reactions |
Each for $873.00
|
![]() |
|||||
Description
Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA. First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.
First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains pre-mixed components to reduce the number of pipetting steps necessary to complete the procedure.
Features of the Maxima H Minus Double-Stranded cDNA Synthesis Kit include:
- Efficient synthesis of full-length double-stranded cDNA
- Fast—procedure completed in less than two hours
- Convenient—pre-mixed components
- Complete—includes all primers, controls and residual RNA removal reagents
Applications
- Full-length double-stranded blunt-end cDNA synthesis for cloning
- Double-stranded cDNA library construction
Specifications
Maxima | |
Maxima H Minus | |
Double Stranded cDNA | |
RNA |
cDNA Synthesis Kit | |
Cloning | |
Separate Components |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.