Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MaxiMix™ I Vortex Mixer

Catalog No. p-4531120
Change view
Click to view available options
Voltage:
120 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
12-815-50 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-815-50 Supplier Thermo Scientific™ Supplier No. M16715Q

Mix quickly and evenly in continuous operation or touch-on mode with this mixer. Excellent for single and multiple tubes and small flasks with closed top.

Mix quickly and evenly in continuous operation or touch-on mode with the Thermo Scientific™ MaxiMix™ I Vortex Mixer. This mixer is ideal for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilutions preparations, precipitated assays, cell suspension vortexing and tissue sample mixing. This mixer is excellent for single and multiple tubes, as well as small flasks with closed tops.

Features:
  • Simultaneously mixes up to four test tubes
  • Push-button top-mounted ON/OFF switch
  • Vary vortex mixing by simply changing pressure of tube against foam rubber top
  • Durable, white plastic case resists acids and alkalis
  • Cast-aluminum base with suction-cup rubber feet provides maximum stability
  • Compact design with 4″ diameter (10.2cm) foam rubber top

Includes:

  • One extra foam top (12-814-1)
  • Three-wire power cord and plug

Recommended for:

  • Enzymatic and RIA assays
  • General test tube mixing
  • Biochemical and precipitated assays
  • Vortexing cell suspensions
  • Mixing tissue samples
  • Viral dilution preparations
TRUSTED_SUSTAINABILITY

Specifications

Speed Range 0 to 3000 rpm
Electrical Requirements 120 V, 50/60 Hz
Voltage 120 V
Amperage 0.5 A
Product Type Mixer
Certifications/Compliance CSA
Frequency 50/60 Hz
Material Cast Aluminum Base, Foam Platform
Dimensions (L x W x H) 5.5 x 5.5 x 4.2 in. (13.9 x 13.9 x 10.6 cm)
Height (English) 4.2 in.
Height (Metric) 10.6 cm
Length (English) 5.5 in.
Length (Metric) 13.9 cm
Width (English) 5.5 in.
Width (Metric) 13.9 cm
Shipping Weight (English) 3.5 lb.
Shipping Weight (Metric) 1.5 kg
Controller Type Push Button On/OFF Switch
Model MaxiMix I Vortex Mixer
No. of Positions 4
Operating Modes Touch, Continuous
Warranty and Service Offering 90 Days (Labor), 1 Year (Parts and Labor)
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.