Learn More
Jackson Safety Maxview™ Series Face Shield - Ratcheting - Chin and Side Guard
The MAXVIEW is a premium face shield that features an oversized window that delivers exceptional optical clarity and panoramic view. An integrated side and chin guard protects against splashes and flying debris while the extended crown protects against hazards from above.
$47.89 - $78.44
Specifications
Height (English) | 13 in. |
---|---|
Height (Metric) | 33.02 cm |
Shape | Contoured |
Width (English) | 9.63 in. |
Width (Metric) | 24.45 cm |
Catalog Number | Mfr. No. | Lens Coating | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Coating | Price | Quantity & Availability | |||||
19-060-842
![]() |
Jackson Safety
14200 |
Uncoated |
Each for $47.89
|
|
|||||
19-060-843
![]() |
Jackson Safety
14201 |
Anti-fog |
Each for $78.44
|
|
|||||
Description
- Extra-large stylish crown and wraparound face shield frame
- High-impact polycarbonate window (with or without anti-fog)
Specifications
13 in. | |
Contoured | |
24.45 cm | |
CE Certified | |
Faceshield | |
9 in. | |
Polycarbonate | |
Polycarbonate | |
None |
33.02 cm | |
9.63 in. | |
ANSI Z87.1 +high impact | |
Clear | |
Black | |
22.86 cm | |
Reusable | |
Polycarbonate |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.