Learn More
Promega Maxwell™ 16 LEV RNA FFPE Kit
Catalog No. pras1260
Click to view available options
Quantity:
48 Preps
The Maxwell 16 LEV RNA FFPE Purification Kit is designed to extract RNA rapidly from FFPE samples using the Maxwell 16 Instrument (Cat.# AS2000 and AS3000).
  • Automated RNA Extraction from 5microm FFPE Sections
  • Maximizes RNA yield. No need to concentrate purified RNA before amplification.
  • Simple method with no organic solvents for deparaffinization
  • Amplification-ready RNA in approximately 60 minutes (after proteinase K digest)
Specifications
Content And Storage 15°C to 30°C
Quantity 48 Preps
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.