Learn More
Promega Maxwell™ RSC RNA FFPE Kit
Catalog No. pras1440
Click to view available options
Quantity:
48 Preps
The Maxwell RSC RNA FFPE Kit is used with the Maxwell RSC Instrument to provide an easy method for efficient, automated purification of RNA from mammalian formalin-fixed, paraffin-embedded (FFPE) tissue samples.
  • Automated RNA Extraction from FFPE Tissue Sections
  • Extracts RNA from 1 to 48 formalin-fixed, paraffin-embedded tissue samples
  • Doesn't use Xylene or organic reagents
  • Extracted RNA is ready for use in downstream amplification assays
Specifications
Content And Storage 15°C to 30°C
Quantity 48 Preps
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.