Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ MAXX Pack Absorbent Pack
Shop All Brady Corporation Products
Click to view available options
Type:
Spill Kit, Oil-only
Universal
Color:
Gray
White
For Use With (Application):
Oil-only spills
Spills
Description
- Economic solution for oil spill containment
- Pack contains 20 full-size, single-ply absorbent pads in a re-sealable zipper bag
- Great for storing around facilities for immediate spill response
Specifications
Specifications
| Absorbancy | 18 gl/gs |
| Color | Gray |
| Capacity (English) | 35 gal. |
| Material | Polypropylene |
| Type | Spill Kit, Oil-only |
| For Use With (Application) | Oil-only spills |
| Packaging Quantity | 5/Pk., 4 Pk./Cs. |
| Dimensions (L x W) | 15 x 19 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction