Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals May-Gruenwald Stain, pure

Catalog No. p-155625
Click to view available options
Quantity:
25 g
100 g

CAS: 524698-43-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material May-Gruenwald Stain
CAS 524698-43-9
MDL Number MFCD00131580
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Lambda Max in 66:33 IMS:water, 655 to 670nm (blue), 515 to 530nm (red)
Grade Pure
Packaging Glass bottle
Color Brown or Green
Quantity 25 g
Physical Form Fine Crystalline Powder
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.