Learn More
Stoelting™ Mayo-Noble Scissors
SDP
Curved or straight scissors
$261.00
Specifications
For Use With (Application) Neuroscience
Length (English) 6.5 in.
View More Specs
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-001-718
SDP
View Documents
Stoelting™
5213668
Straight
Each for $261.00
Only null left
Add to Cart
10-001-719
SDP
View Documents
Stoelting™
5213669
Curved
Each for $261.00
Only null left
Add to Cart
Description
Description
  • Straight-bladed style is designed for cutting body tissues near the surface of a wound.
  • Curved-bladed styles allow deeper penetration into the wound and are also used to cut thick tissues
Specifications
Specifications
Neuroscience
6.5 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.