Learn More
Stoelting™ Mayo Scissors
SDP
$162.60 - $503.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 15
Catalog Number Mfr. No. Length (English) Model Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Model Price Quantity & Availability  
10-001-703
SDP
View Documents
Stoelting™
5213653
5.5 in. Curved, Surecut
Each for $365.50
Only null left
Add to Cart
10-001-701
SDP
View Documents
Stoelting™
5213651
5.5 in. Straight, Surecut
Each for $392.50
Only null left
Add to Cart
10-000-544
SDP
View Documents
Stoelting™
5213661P
5.5 in. Curved, Tungsten carbide
Each for $270.00
Only null left
Add to Cart
10-000-543
SDP
View Documents
Stoelting™
5213660P
5.5 in. Straight, Tungsten carbide
Each for $285.00
Only null left
Add to Cart
10-000-541
SDP
View Documents
Stoelting™
5213652P
5.75 in. Curved
Each for $162.60
Only null left
Add to Cart
10-000-540
SDP
View Documents
Stoelting™
5213650P
5.75 in. Straight
Each for $162.60
Only null left
Add to Cart
10-001-707
SDP
View Documents
Stoelting™
5213657
6.75 in. Curved
Each for $242.00
Only null left
Add to Cart
10-001-713
SDP
View Documents
Stoelting™
5213663
6.75 in. Curved Carbide
Each for $453.00
Only null left
Add to Cart
10-001-720
SDP
View Documents
Stoelting™
5213670
6.75 in. Curved, Surecut
Each for $365.50
Only null left
Add to Cart
10-001-704
SDP
View Documents
Stoelting™
5213654
6.75 in. Straight
Each for $233.50
Only null left
Add to Cart
10-001-712
SDP
View Documents
Stoelting™
5213662
6.75 in. Straight Carbide
Each for $431.00
Only null left
Add to Cart
10-001-706
SDP
View Documents
Stoelting™
5213656
6.75 in. Straight, Surecut
Each for $365.50
Only null left
Add to Cart
10-000-542
SDP
View Documents
Stoelting™
5213655P
6.75 in. Straight, Left-handed
Each for $230.50
Only null left
Add to Cart
10-001-709
SDP
View Documents
Stoelting™
5213659
9 in. Curved, Surecut
Each for $503.00
Only null left
Add to Cart
10-001-708
SDP
View Documents
Stoelting™
5213658
9 in. Straight, Surecut
Each for $503.00
Only null left
Add to Cart
Description
Description
  • Straight-bladed style is designed for cutting body tissues near the surface of a wound.
  • Curved-bladed styles allow deeper penetration into the wound and are also used to cut thick tissues
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.