Learn More
Stoelting™ Mayo-Stille Scissors
SDP
Straight and curved scissors
$198.30 - $217.50
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 4
Catalog Number Mfr. No. Length (English) Model Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Model Price Quantity & Availability  
10-001-714
SDP
View Documents
Stoelting™
5213664
5.5 in. Straight
Each for $198.30
Only null left
Add to Cart
10-001-715
SDP
View Documents
Stoelting™
5213665
5.5 in. Curved
Each for $198.30
Only null left
Add to Cart
10-001-716
SDP
View Documents
Stoelting™
5213666
6.75 in. Straight
Each for $217.50
Only null left
Add to Cart
10-001-717
SDP
View Documents
Stoelting™
5213667
6.75 in. Curved
Each for $217.50
Only null left
Add to Cart
Description
Description
  • Straight-blade style is designed for cutting body tissues near the surface of a wound.
  • Curved-blade style allows deeper penetration into the wound than the type with straight blades and is used to cut thick tissues
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.