Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maz Monoclonal Antibody (133), Active Motif™
SDP

Catalog No. 501992977
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2977 100 μg
50-199-2978 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2977 Supplier Active Motif Supplier No. 39935
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors.

Specifications

Antigen Maz
Applications Western Blot
Classification Monoclonal
Clone 133
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene MAZ
Gene Accession No. P56270
Gene Alias MAZ; MAZI; MYC associated zinc finger protein; Myc-associated zinc finger protein; MYC-associated zinc finger protein (purine-binding transcription factor); PUR1; Pur-1; Purine-binding transcription factor; SAF-1; SAF-2; SAF-3; serum amyloid A activating factor 1; serum amyloid A activating factor 2; Serum amyloid A-activating factor-1; Transcription factor Zif87; ZF87; Zif87; Zinc finger protein 801; zinc-finger protein, 87 kilodaltons; ZNF801
Gene Symbols MAZ
Host Species Mouse
Immunogen This MAZ antibody was raised against recombinant human MAZ protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4150
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.