Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588863 Supplier Thermo Scientific Supplier No. PA588863
Add to Cart
Add to Cart

MAZ Polyclonal Antibody

MAZ Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAZ
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MAZ
Gene Accession No. P56270
Gene Alias LOW QUALITY PROTEIN: myc-associated zinc finger protein; Maz; MAZI; MYC associated zinc finger protein; myc-associated zinc finger protein; MYC-associated zinc finger protein (purine-binding transcription factor); PUR1; Pur-1; purine-binding transcription factor; SAF-1; SAF-2; SAF-3; serum amyloid A activating factor 1; serum amyloid A activating factor 2; serum amyloid A-activating factor-1; Transcription factor Zif87; ZF87; Zif87; Zinc finger protein 801; zinc-finger protein, 87 kilodaltons; ZNF801
Gene Symbols MAZ
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 270-420 of human MAZ (NP_0010360041).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4150
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.