Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106686 Supplier Invitrogen™ Supplier No. PA5106686
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAZ.

May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors. Regulates inflammation-induced expression of serum amyloid A proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAZ
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAZ
Gene Accession No. P56270, P56671
Gene Alias LOW QUALITY PROTEIN: myc-associated zinc finger protein; Maz; MAZI; MYC associated zinc finger protein; myc-associated zinc finger protein; MYC-associated zinc finger protein (purine-binding transcription factor); PUR1; Pur-1; purine-binding transcription factor; SAF-1; SAF-2; SAF-3; serum amyloid A activating factor 1; serum amyloid A activating factor 2; serum amyloid A-activating factor-1; Transcription factor Zif87; ZF87; Zif87; Zinc finger protein 801; zinc-finger protein, 87 kilodaltons; ZNF801
Gene Symbols MAZ
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAZ(Accession P56270), corresponding to amino acid residues R51-A101.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17188, 4150, 691842
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.