Learn More
MAZ/SAF-1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301652AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors.Specifications
| MAZ/SAF-1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P56270, P56671 | |
| Maz | |
| Between 427 and 477 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| MAZ | |
| PUR1, Pur-1, SAF-1, SAF-2, SAF-3, ZF87, ZNF801, Zif87, MAZI, myc-associated zinc finger protein, purine-binding transcription factor, serum amyloid A activating factor 1, serum amyloid A activating factor 2, transcription factor Zif87, zinc finger protein 801, zinc-finger protein, 87 kilodaltons | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17188, 4150 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.