Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MB Mouse anti-Human, Mouse, Rat, Unconjugated, Clone: MB/2105, Abnova™

Catalog No. 89337793 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-793 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-793 Supplier Abnova Corporation Supplier No. MAB21302
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MB Monoclonal antibody specifically detects MB in Human, Mouse, Rat samples. It is validated for IHC-P.

Specifications

Antigen MB
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MB/2105
Conjugate Unconjugated
Description myoglobin
Gene MB
Gene Alias MGC13548, PVALB
Host Species Mouse
Immunogen Recombinant protein corresponding to full-length human MB
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4151
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.