Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MB67 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120678 Supplier Invitrogen™ Supplier No. PA5120678
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, Mouse liver, Rat liver. The target is usually found in the following locations: Cytoplasm, Nucleus, cytoskeleton. Immunogen sequence: PVQLSKEQEE LIRTLLGAHT RHMGTMFEQF VQFRPPAHLF IHHQPLPTLA PVLPLVTHFA DINTFMVLQV IKFTKDLPVF RSLPIEDQIS LLKGAAVEIC HIVLNTTFCL QTQNFLCGPL RYTIEDGARV SPTVGFQVEF LELLFHFHGT LRKLQLQEPE YVLLAAMALF SPDRPGVTQR DEIDQLQEEM ALTLQSYIKG QQRRPRDRFL YAKLLGLLAE LRSINEAYGY QIQHIQGLSA MMPLLQEICS.

This gene encodes a member of the nuclear receptor superfamily, and is a key regulator of xenobiotic and endobiotic metabolism. The protein binds to DNA as a monomer or a heterodimer with the retinoid X receptor and regulates the transcription of target genes involved in drug metabolism and bilirubin clearance, such as cytochrome P450 family members. Unlike most nuclear receptors, this transcriptional regulator is constitutively active in the absence of ligand but is regulated by both agonists and inverse agonists.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MB67
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene NR1I3
Gene Accession No. O35627, Q14994, Q9QUS1
Gene Alias AA209988; AI551208; Car; CAR1; CAR-beta; Care2; constitutive activator of retinoid response; constitutive active receptor; constitutive active response; constitutive androstane nuclear receptor variant 2; constitutive androstane nuclear receptor variant 3; constitutive androstane nuclear receptor variant 4; constitutive androstane nuclear receptor variant 5; constitutive androstane receptor; constitutive androstane receptor NR1I3; constitutive androstane receptor protein variant 1; constitutive androstane receptor protein variant 2; constitutive androstane receptor protein variant 3; constitutive androstane receptor protein variant 4; constitutive androstane receptor protein variant 5; constitutively transactivates RAREs; ESTM32; MB67; MGC150433; MGC97144; MGC97209; NR1I3; nuclear receptor; nuclear receptor constitutive active receptor; nuclear receptor subfamily 1 group I member 3; nuclear receptor subfamily 1, group I, member 3; orphan nuclear hormone receptor; orphan nuclear receptor MB67; strain Fischer nuclear receptor (CAR); Unknown (protein for MGC:137128)
Gene Symbols NR1I3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 103-352 of human NR1I3 (NP_001070948.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12355, 65035, 9970
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.