Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mBaa Mouse anti-Tag, Magnetic beads, Clone: OTI2E5, liquid, TrueMAB™
SDP

Catalog No. 501671180
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1180 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1180 Supplier OriGene Supplier No. TA183032
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

mBaa fluorescent protein.

Specifications

Antigen mBaa
Applications Immunoprecipitation
Classification Monoclonal
Clone OTI2E5
Formulation PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide
Gene Alias mBaa
Host Species Mouse
Immunogen Recombit protein of mBaa expressed in HEK293 cells.
Purification Method Affinity Chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.