Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD, 98%

Catalog No. p-4509618
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413540010 1 g Glass bottle
AC413540050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413540010 Supplier Thermo Scientific Chemicals Supplier No. 413540010
Only null left
Add to Cart

CAS: 33984-50-8 | C14H12N4O4 | 300.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33984-50-8
Molecular Formula C14H12N4O4
Molecular Weight (g/mol) 300.26
InChI Key IENONFJSMWUIQQ-UHFFFAOYSA-N
Synonym 4-4-methoxybenzylamino-7-nitrobenzofurazan, 4-4-methoxybenzylamino-7-nitro-2,1,3-benzoxadiazole, mbd, 7-p-methoxybenzylamino-4-nitrobenz-2-oxa-1,3-diazole, 2,1,3-benzoxadiazol-4-amine, n-4-methoxyphenyl methyl-7-nitro, benzofurazan, 4-p-methoxybenzyl amino-7-nitro, n-4-methoxyphenyl methyl-7-nitro-2,1,3-benzoxadiazol-4-amine
PubChem CID 99555
IUPAC Name N-[(4-methoxyphenyl)methyl]-4-nitro-2,1,3-benzoxadiazol-7-amine
SMILES COC1=CC=C(C=C1)CNC2=CC=C(C3=NON=C23)[N+](=O)[O-]

Specifications

Melting Point 176.0°C to 180.0°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 300.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material MBD
EINECSNumber 251-775-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.