Learn More
Description
Specifications
Specifications
| Antigen | MBD1 |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown |
| Classification | Monoclonal |
| Clone | 100B272.1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2-4 ug/ml, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 25798578), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24464130), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 15327775), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 24464130), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated |
| Gene Accession No. | Q9UIS9 |
| Gene Alias | CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription |
| Gene Symbols | MBD1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
