Learn More
MBD1 Mouse anti-Human, Clone: OTI2D7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811768

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Methyl-CpG-binding domain protein 1 (MBD1) is a member of a family of mammalian proteins that specifically bind methylated regions of DNA. It is a transcriptional repressor that binds CpG islands in promoters where the DNA is methylated at position 5 of cytosine within CpG dinucleotides. Binding is abolished by the presence of 7-mG that is produced by DNA damage by methylmethanesulfonate (MMS). MBD1 acts as a transcriptional repressor and plays a role in gene silencing by recruiting AFT7IP, which in turn recruits factors such as the histone methyltransferase SETDB1. Isoform 1 and isoform 2 can also repress transcription form unmethylated promoters.Specifications
MBD1 | |
Monoclonal | |
Unconjugated | |
MBD1 | |
CXXC3, PCM1, RFT | |
Mouse | |
Affinity Chromatography | |
RUO | |
4152 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2D7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UIS9 | |
MBD1 | |
Human recombit protein fragment corresponding to amino acids 222-556 of human MBD1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.