Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA129797
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129797 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129797 Supplier Invitrogen™ Supplier No. PA129797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-29797 detects MBD1 from human samples.

Methyl-CpG-binding domain protein 1 (MBD1) is a member of a family of mammalian proteins that specifically bind methylated regions of DNA. It is a transcriptional repressor that binds CpG islands in promoters where the DNA is methylated at position 5 of cytosine within CpG dinucleotides. Binding is abolished by the presence of 7-mG that is produced by DNA damage by methylmethanesulfonate (MMS). MBD1 acts as a transcriptional repressor and plays a role in gene silencing by recruiting AFT7IP, which in turn recruits factors such as the histone methyltransferase SETDB1. Isoform 1 and isoform 2 can also repress transcription from unmethylated promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MBD1
Gene Accession No. Q9UIS9
Gene Alias CXXC3; CXXC-type zinc finger protein 3; Mbd1; methyl-CpG binding domain protein 1; methyl-CpG binding domain protein 1 isoform PCM1; methyl-CpG-binding domain protein 1; Methyl-CpG-binding protein MBD1; PCM1; Protein containing methyl-CpG-binding domain 1; RFT; the regulator of fibroblast growth factor 2 (FGF-2) transcription
Gene Symbols MBD1
Host Species Rabbit
Immunogen Mixture of synthetic peptides corresponding to amino acids 98-113 (RQVGPQSGEVRKEAPR) and 391-405 (SESEDGAGSPPPYRR) of human MBD1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4152
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.